Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hecw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001883.3 | NP_001001883.1 | 2 | 342 | ||
NM_172655.3 | NP_766243.2 | 2 | 342 | |||
GenBank mRNA | AB182244.1 | - | 1 | 248 | ||
AK042922.1 | - | 2 | 343 | |||
AK047602.1 | - | 2 | 477 | |||
BC107219.2 | - | 2 | 327 | |||
BC134353.1 | - | 2 | 326 | |||
BC148188.2 | - | 2 | 325 |