Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam163a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177838.3 | NP_808506.1 | 1077 | |||
GenBank mRNA | AK039197.1 | - | 1078 | |||
BC116972.1 | - | 2 | 746 | 40126291 | ||
BC116974.1 | - | 2 | 746 | 40126293 |