Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301370.1 | NP_001288299.1 | 6 | 1173 | ||
NM_181546.3 | NP_853524.1 | 6 | 1173 | |||
GenBank mRNA | AB102947.1 | - | 6 | 1096 | ||
AK051175.1 | - | 3 | 2847 | |||
BC141262.1 | - | 6 | 1115 | 9055874 | ||
BC145602.1 | - | 6 | 1115 | 9054168 |