Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nostrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181547.3 | NP_853525.3 | 5 | 372 | ||
GenBank mRNA | AJ564264.1 | - | 5 | 282 | ||
AK052926.1 | - | 5 | 364 | |||
AY280963.1 | - | 5 | 325 | |||
BC069942.1 | - | 5 | 349 | 30443942 | ||
BY061165.1 | - | 5 | 372 |