Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290782.1 | NP_001277711.1 | 15 | 1632 | ||
NM_183220.3 | NP_899043.1 | 15 | 1687 | |||
GenBank mRNA | AK011867.1 | - | 15 | 1514 | ||
BC039569.1 | - | 15 | 1664 | 1449080 |