Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163940.1 | NP_001157412.1 | 14 | 1841 | ||
NM_002863.4 | NP_002854.3 | 15 | 1943 | |||
GenBank mRNA | AF046785.1 | - | 15 | 1816 | ||
AF066858.1 | - | 15 | 1816 | |||
AK225904.1 | - | 15 | 1896 | |||
AK300580.1 | - | 14 | 1794 | |||
AK309333.1 | - | 15 | 1869 | |||
AK313206.1 | - | 15 | 1857 | |||
BC009895.2 | - | 15 | 1886 | 2967205 | ||
BC082229.1 | - | 15 | 1887 | 4053354 | ||
BC095850.1 | - | 15 | 1903 | 30343100 | ||
BC110791.1 | - | 15 | 1885 | 6051020 | ||
M14636.1 | - | 15 | 1929 | |||
M36807.1 | - | 4 | 373 | |||
Y15233.1 | - | 15 | 1867 |