Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025662.5 | NP_079938.1 | 4 | 384 | ||
NM_178016.3 | NP_821135.1 | 4 | 384 | |||
GenBank mRNA | AK013853.1 | - | 4 | 381 | ||
AK043033.1 | - | 373 | ||||
AK046246.1 | - | 4 | 372 | |||
AK077893.1 | - | 4 | 385 | |||
AK082806.1 | - | 4 | 377 | |||
AK160801.1 | - | 4 | 379 | |||
AK168666.1 | - | 366 | ||||
BC023256.1 | - | 4 | 366 | |||
BC047203.1 | - | 1 | 34 | |||
BC060175.1 | - | 4 | 370 | 6827521 |