Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177867.3 | NP_808535.1 | 3 | 417 | ||
GenBank mRNA | AK077155.1 | - | 3 | 418 | ||
BC112427.1 | - | 2 | 126 | 40062127 | ||
BC113132.1 | - | 2 | 126 | 40062125 |