Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem120b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039723.2 | NP_001034812.1 | 6 | 666 | ||
GenBank mRNA | AK132283.1 | - | 6 | 635 | ||
AK158123.1 | - | 6 | 625 | |||
AK172110.1 | - | 6 | 606 | |||
BC022593.1 | - | 6 | 617 | 4038363 | ||
BC115785.1 | - | 5 | 510 | 40104955 | ||
BC118014.1 | - | 6 | 627 | 40104954 |