Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177879.5 | NP_808547.3 | 15, 16 | 2545 | ||
GenBank mRNA | AK147578.1 | - | 15, 16 | 2590 | ||
AY353236.1 | - | 11, 12 | 1695 | |||
BC060237.1 | - | 15, 16 | 2129 | |||
BC138453.1 | - | 11, 12 | 1566 | 8861473 |