Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198884.1 | NP_942585.1 | 18 | 2612 | ||
GenBank mRNA | AB114826.1 | - | 18 | 2612 | ||
AK082874.1 | - | 4 | 330 | |||
AK143898.1 | - | 18 | 2528 | |||
BC139798.1 | - | 18 | 2626 | 40060840 |