Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198631.2 | NP_941033.2 | 2 | 321 | ||
GenBank mRNA | AK138078.1 | - | 1 | 2139 | ||
AK151148.1 | - | 3 | 373 | |||
AK151306.1 | - | 3 | 373 | |||
BC057627.1 | - | 2 | 266 | 6412890 | ||
BY167917.1 | - | 3 | 370 | |||
CJ101946.1 | - | 2 | 326 |