Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252519.1 | NP_001239448.1 | 6 | 574 | ||
NM_177889.5 | NP_808557.2 | 7 | 664 | |||
GenBank mRNA | AK129467.1 | - | 2 | 2075 | ||
AK137630.1 | - | 288 | ||||
BC060530.1 | - | 6 | 591 | 5684819 | ||
CD806198.1 | - | 5 | 525 | |||
U29496.1 | - | 3 | 182 |