Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177893.3 | NP_808561.2 | 3 | 1684 | ||
GenBank mRNA | AK077918.1 | - | 3 | 1675 | ||
AK173087.1 | - | 2 | 1142 | |||
BC116270.2 | - | 1 | 2408 | |||
BC116271.2 | - | 1 | 2406 |