Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039514.1 | NP_001034603.1 | 9 | 1152 | ||
GenBank mRNA | AK159104.1 | - | 9 | 1153 | ||
AK212076.1 | - | 2 | 72 | |||
BB763715.1 | - | 4 | 345 | |||
BC039963.1 | - | 7 | 938 | 4240646 | ||
BC061234.1 | - | 8 | 991 |