Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178798.3 | NP_848913.1 | 10 | 1608 | ||
GenBank mRNA | AK030762.1 | - | 11 | 1410 | ||
AK046548.1 | - | 9 | 2275 | |||
AK049478.1 | - | 10 | 1608 | |||
AK053899.1 | - | 10 | 1617 | |||
AK088592.1 | - | 10 | 1525 | |||
AK129099.1 | - | 7 | 3004 | |||
AK157127.1 | - | 10 | 1747 | |||
AK157379.1 | - | 9 | 2277 | |||
AK164666.1 | - | 10 | 1609 | |||
AK165629.1 | - | 10 | 1625 | |||
AK165979.1 | - | 10 | 1627 | |||
AK194286.1 | - | 2 | 70 | |||
AK205341.1 | - | 2 | 70 | |||
AK209516.1 | - | 2 | 70 | |||
AK210535.1 | - | 2 | 70 | |||
AK218208.1 | - | 2 | 70 | |||
BC038404.1 | - | 9 | 1543 | 4949962 | ||
BC042898.1 | - | 7 | 2140 |