Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014900.2 | NP_001014900.2 | 6 | 933 | ||
GenBank mRNA | AY959941.1 | - | 6 | 744 | ||
BC147552.1 | - | 6 | 843 | 9056543 | ||
BC147553.1 | - | 6 | 843 | 9056544 | ||
BC147848.1 | - | 6 | 933 | 9087565 | ||
BC147851.1 | - | 6 | 933 | 9087568 |