Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175380.5 | NP_780589.3 | 1, 2 | 328 | ||
GenBank mRNA | AK050572.1 | - | 1, 2 | 221 | ||
AK075845.1 | - | 1, 2 | 170 | |||
AK145475.1 | - | 1, 2 | 183 | |||
AK166662.1 | - | 1, 2 | 180 | |||
AK172886.1 | - | 1, 2 | 273 | |||
BC037729.1 | - | 1, 2 | 191 | |||
BC138976.1 | - | 1, 2 | 156 | 8861998 | ||
BC138980.1 | - | 1, 2 | 156 | 8862002 |