Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177381.3 | NP_796355.2 | 12 | 1343 | ||
GenBank mRNA | AK140362.1 | - | 12 | 1343 | ||
BC038030.1 | - | 13 | 1354 | 4020725 | ||
BC048005.1 | - | 8 | 698 |