Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045528.1 | NP_001038993.1 | 17 | 1786 | ||
GenBank mRNA | AK049459.1 | - | 17 | 1731 | ||
AK049509.1 | - | 17 | 1786 | |||
AK082565.1 | - | 17 | 1735 | |||
AK085087.1 | - | 17 | 1777 | |||
AK122343.1 | - | 10 | 1025 | |||
AK161705.1 | - | 17 | 1716 | |||
BC034098.1 | - | 7 | 723 | |||
BC060651.1 | - | 17 | 1700 | |||
BC137706.1 | - | 17 | 1720 | 8860725 |