Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289666.1 | NP_001276595.1 | 11 | 673 | ||
NM_001289667.1 | NP_001276596.1 | 9 | 513 | |||
NM_178801.5 | NP_848916.2 | 11 | 673 | |||
GenBank mRNA | AK028973.1 | - | 223 | |||
AK031517.1 | - | 12 | 759 | |||
AK038419.1 | - | 11 | 651 | |||
AK038472.1 | - | 11 | 618 | |||
AK080479.1 | - | 2 | 2324 | |||
AK144963.1 | - | 11 | 675 | |||
BC094382.1 | - | 11 | 589 | 30605060 | ||
BC137621.1 | - | 11 | 603 | 8860640 | ||
BC144788.1 | - | 9 | 444 |