Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177392.3 | NP_796366.1 | 2, 3 | 146 | ||
GenBank mRNA | AK043094.1 | - | 2, 3 | 147 |