Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290776.1 | NP_001277705.1 | 6 | 674 | ||
NM_001290777.1 | NP_001277706.1 | 6 | 674 | |||
NM_001290778.1 | NP_001277707.1 | 5 | 821 | |||
NM_178935.5 | NP_849266.1 | 5 | 821 | |||
GenBank mRNA | AK030100.1 | - | 5 | 768 | ||
AK031023.1 | - | 4 | 1493 | |||
AK031783.1 | - | 5 | 735 | |||
AK044130.1 | - | 5 | 770 | |||
AK078477.1 | - | 6 | 621 | |||
AK134415.1 | - | 5 | 1497 | |||
AK161561.1 | - | 6 | 583 | |||
AK162263.1 | - | 5 | 685 | |||
AK166902.1 | - | 5 | 1346 | |||
BC059099.1 | - | 5 | 751 | |||
BC080778.1 | - | 5 | 733 | |||
BC117898.1 | - | 6 | 615 | 40096960 | ||
BC117899.1 | - | 6 | 615 | 40096967 | ||
DQ316984.1 | - | 5 | 821 | |||
DQ320011.1 | - | 6 | 674 |