Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181348.4 | NP_851993.3 | 8 | 1365 | ||
GenBank mRNA | AK039241.1 | - | 8 | 1366 |