Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198117.2 | NP_932785.1 | 1 | 866 | ||
GenBank mRNA | AY013760.1 | - | 1 | 753 | ||
BC030897.1 | - | 1 | 866 | |||
BC141390.1 | - | 1 | 847 | 9056003 |