Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181549.3 | NP_853527.2 | 2 | 229 | ||
GenBank mRNA | AK132888.1 | - | 4 | 1036 | ||
AY223868.1 | - | 437 | ||||
AY223869.1 | - | 408 | ||||
AY223870.1 | - | 412 | ||||
AY223871.1 | - | 318 | ||||
AY223872.1 | - | 383 | ||||
AY223873.1 | - | 289 |