Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181683.2 | NP_859011.1 | 2 | 213 | ||
GenBank mRNA | AJ489588.1 | - | 2 | 213 | ||
AJ578470.1 | - | 2 | 131 |