Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181820.2 | NP_861541.2 | 7 | 1139 | ||
GenBank mRNA | AK050105.1 | - | 6 | 1034 | ||
AY236493.1 | - | 7 | 1102 | |||
AY263162.1 | - | 7 | 1110 | |||
BC040466.1 | - | 7 | 1051 | |||
BC055939.1 | - | 6 | 868 | |||
BC106105.1 | - | 6 | 1631 |