Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183036.1 | NP_898857.1 | 2 | 107 | ||
GenBank mRNA | AJ575424.1 | - | 2 | 144 | ||
AJ578471.1 | - | 2 | 107 | |||
BC125388.1 | - | 2 | 151 | 40129903 | ||
BC125390.1 | - | 2 | 151 | 40129905 |