Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183038.2 | NP_898859.2 | 2 | 152 | ||
GenBank mRNA | AJ578472.1 | - | 2 | 152 |