Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278622.1 | NP_001265551.1 | 16 | 2173 | ||
NM_002873.1 | NP_002864.1 | 15 | 2278 | |||
NM_133338.2 | NP_579916.1 | 17 | 2174 | |||
NM_133339.2 | NP_579917.1 | 14 | 1731 | |||
NM_133340.2 | NP_579918.1 | 14 | 1764 | |||
NM_133341.2 | NP_579919.1 | 14 | 1690 | |||
NM_133342.2 | NP_579920.1 | 16 | 2151 | |||
NM_133343.1 | NP_579921.1 | 16 | 2116 | |||
NM_133344.2 | NP_579922.1 | 15 | 1933 | |||
XM_017009679.1 | XP_016865168.1 | 16 | 1993 | |||
XM_017009680.1 | XP_016865169.1 | 15 | 2047 | |||
XM_017009681.1 | XP_016865170.1 | 15 | 1905 | |||
GenBank mRNA | AF017748.1 | - | 15 | 1903 | ||
AF076838.1 | - | 15 | 2278 | |||
AF085736.2 | - | 14 | 1731 | |||
AF098533.1 | - | 14 | 1710 | |||
AF098534.1 | - | 14 | 1636 | |||
AF126424.1 | - | 17 | 2173 | |||
AJ001642.1 | - | 15 | 1942 | |||
AJ004977.1 | - | 15 | 1938 | |||
AK292487.1 | - | 16 | 2151 | |||
AL122068.1 | - | 16 | 2116 | |||
BX537441.1 | - | 16 | 2173 |