Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025685.3 | NP_079961.3 | 60 | 5660 | ||
GenBank mRNA | AK017766.1 | - | 1 | 363 | ||
AK053733.1 | - | 39 | 2512 | |||
AK173283.1 | - | 36 | 2284 | |||
AY167568.2 | - | 60 | 5246 | |||
BC038041.1 | - | 13 | 894 | |||
BC039222.1 | - | 57 | 3585 | |||
BC049173.1 | - | 57 | 3573 |