Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002842.2 | NP_001002842.2 | 2 | 900 | ||
GenBank mRNA | AK169990.1 | - | 2 | 890 | ||
AY665714.1 | - | 2 | 816 | |||
BC141100.1 | - | 2 | 805 | 9055712 |