Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194357.1 | NP_919338.2 | 5 | 606 | ||
GenBank mRNA | BC048084.1 | - | 5 | 606 | 6312261 |