Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147155.2 | NP_671511.2 | 3 | 196 | ||
GenBank mRNA | AF314818.1 | - | 3 | 196 | ||
AV444953.1 | - | 3 | 240 | |||
BC119550.1 | - | 3 | 196 | |||
BC119551.1 | - | 3 | 196 | |||
BC139797.1 | - | 3 | 194 | |||
BQ555561.1 | - | 2 | 71 |
Gene Symbol: Tagap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145968.2 | NP_666080.1 | 9 | 959 | ||
GenBank mRNA | BC030886.1 | - | 9 | 939 | 4483459 | |
BC039780.1 | - | 9 | 950 | 4483459 | ||
BC150728.1 | - | 9 | 904 | 9087639 |