Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033434.2 | NP_001028606.2 | 6 | 2186 | ||
GenBank mRNA | AK029868.1 | - | 6 | 2186 | ||
JQ790768.1 | - | 6 | 496 | |||
JQ790769.1 | - | 6 | 496 | |||
JQ790770.1 | - | 6 | 496 | |||
JQ790771.1 | - | 6 | 496 | |||
JQ790772.1 | - | 6 | 496 | |||
JQ790773.1 | - | 6 | 496 | |||
JQ790774.1 | - | 6 | 496 | |||
JQ790775.1 | - | 6 | 496 | |||
JQ790776.1 | - | 6 | 496 | |||
JQ790777.1 | - | 6 | 496 | |||
JQ790778.1 | - | 6 | 496 | |||
JQ790785.1 | - | 5 | 370 | |||
JQ790786.1 | - | 4 | 352 | |||
JQ790787.1 | - | 4 | 352 | |||
JQ790788.1 | - | 6 | 496 |