Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Milr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033435.3 | NP_001028607.1 | 1 | 100 | ||
NM_001271372.1 | NP_001258301.1 | 1 | 100 | |||
NM_001271373.1 | NP_001258302.1 | 1 | 100 | |||
NM_001271374.1 | NP_001258303.1 | 1 | 100 | |||
NM_001271375.1 | NP_001258304.1 | 1 | 100 | |||
GenBank mRNA | AB542953.1 | - | 1 | 58 | ||
AK136488.1 | - | 1 | 60 | |||
AK137500.1 | - | 1 | 59 | |||
AK151814.1 | - | 1 | 58 | |||
AK170947.1 | - | 1 | 59 | |||
AK171381.1 | - | 1 | 59 | |||
BC139224.1 | - | 1 | 86 | 8862246 | ||
BC139225.1 | - | 1 | 86 | 8862247 | ||
BC145580.1 | - | 1 | 86 | 9054146 | ||
BC145582.1 | - | 1 | 86 | 9054148 | ||
BY197245.1 | - | 1 | 104 |