Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201357.2 | NP_958745.2 | 8 | 1046 | ||
GenBank mRNA | AK150308.1 | - | 8 | 1047 | ||
AK151830.1 | - | 8 | 1047 | |||
BC031458.1 | - | 8 | 1009 | 4191954 |