Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033436.3 | NP_001028608.3 | 8 | 1663 | ||
GenBank mRNA | AK036472.1 | - | 8 | 1613 | ||
AK156016.1 | - | 8 | 1627 | |||
AK158590.1 | - | 5 | 1808 | |||
BC147479.1 | - | 8 | 1578 | 9056221 | ||
BC147483.1 | - | 8 | 1578 | 9056229 |