Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033770.1 | NP_001028942.1 | 1 | 111 | ||
GenBank mRNA | AK131725.1 | - | 1 | 111 | ||
BC147620.1 | - | 1 | 101 | 9056610 |
Gene Symbol: Gm8765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244649.1 | NP_001231578.1 | 1 | 94 | ||
GenBank mRNA | BC172081.1 | - | 1 | 94 | 9054367 |
Gene Symbol: Gm906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033438.2 | NP_001028610.1 | 1 | 111 | ||
GenBank mRNA | AK133043.1 | - | 1 | 112 | ||
BC147515.1 | - | 1 | 94 | 9056507 |