Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033441.3 | NP_001028613.1 | 3 | 1375 | ||
GenBank mRNA | AK143038.1 | - | 3 | 1381 | ||
AK147821.1 | - | 3 | 1376 | |||
AK153580.1 | - | 3 | 1206 | |||
AK166344.1 | - | 3 | 1377 | |||
AK170450.1 | - | 3 | 1377 | |||
AK171179.1 | - | 3 | 1188 |