Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145888.1 | NP_001139360.1 | 3 | 405 | ||
NM_198644.2 | NP_941046.1 | 3 | 405 | |||
GenBank mRNA | BC053930.1 | - | 3 | 396 | 6389756 | |
BY257018.1 | - | 3 | 407 | |||
EU221277.1 | - | 3 | 235 |