Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031621.2 | NP_001026792.2 | 26 | 4444 | ||
GenBank mRNA | AB112584.1 | - | 26 | 4444 |