Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002880.3 | NP_002871.1 | 5 | 907 | ||
XM_005265355.2 | XP_005265412.1 | 5 | 635 | |||
XM_005265357.2 | XP_005265414.1 | 5 | 823 | |||
XM_005265358.4 | XP_005265415.1 | 4 | 789 | |||
XM_005265359.4 | XP_005265416.1 | 4 | 789 | |||
XM_011533974.2 | XP_011532276.1 | 6 | 950 | |||
XM_011533975.2 | XP_011532277.1 | 5 | 839 | |||
XM_017006966.1 | XP_016862455.1 | 5 | 611 | |||
GenBank mRNA | AK226028.1 | - | 5 | 823 | ||
AK303561.1 | - | 4 | 710 | |||
AK303920.1 | - | 1 | 83 | |||
AK312248.1 | - | 5 | 801 | |||
BC018119.2 | - | 5 | 821 | 3904404 | ||
KU178297.1 | - | 4 | 492 | |||
KU178298.1 | - | 4 | 492 | |||
KU178299.1 | - | 4 | 492 | |||
X03484.1 | - | 5 | 621 |