Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000536.3 | NP_000527.2 | 2 | 279 | ||
NM_001243785.1 | NP_001230714.1 | 3 | 474 | |||
NM_001243786.1 | NP_001230715.1 | 3 | 668 | |||
GenBank mRNA | AK292664.1 | - | 2 | 229 | ||
BC022397.1 | - | 2 | 236 | 4656700 | ||
BM457214.1 | - | 2 | 250 | |||
DB124002.1 | - | 3 | 431 | |||
DB142398.1 | - | 2 | 271 |