Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167949.2 | NP_001161421.1 | 17 | 3360 | ||
NM_213616.4 | NP_998781.1 | 17 | 3360 | |||
GenBank mRNA | AY560895.1 | - | 17 | 2894 | ||
BC109172.1 | - | 17 | 2894 | 40048499 | ||
BC109173.1 | - | 17 | 2894 | 40048500 | ||
EU493094.2 | - | 17 | 3064 | |||
EU493095.2 | - | 17 | 3064 | |||
JF683384.1 | - | 17 | 3064 |