Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287179.2 | NP_001274108.1 | 24, 25 | 4286 | ||
GenBank mRNA | AB731728.1 | - | 24, 25 | 4176 | ||
BC054123.1 | - | 12, 13 | 1574 | |||
BC158132.1 | - | 24, 25 | 4199 | 9007399 |