Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198653.2 | NP_941055.1 | 9 | 1220 | ||
GenBank mRNA | AK036182.1 | - | 9 | 1188 | ||
AK043590.1 | - | 9 | 1222 | |||
AK043729.1 | - | 9 | 1198 | |||
AK048653.1 | - | 9 | 1221 | |||
AK050358.1 | - | 9 | 1203 | |||
AK051778.1 | - | 9 | 1181 | |||
AK078183.1 | - | 9 | 1222 | |||
AK085714.1 | - | 9 | 1217 | |||
AK088965.1 | - | 9 | 1181 | |||
AK166832.1 | - | 9 | 1203 | |||
BC037123.1 | - | 4 | 280 | |||
BC052403.1 | - | 9 | 1197 | 5715723 |