Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198653.2 | NP_941055.1 | 19, 20 | 2531 | ||
GenBank mRNA | AK036182.1 | - | 19, 20 | 2499 | ||
AK043590.1 | - | 19, 20 | 2533 | |||
AK048653.1 | - | 19, 20 | 2532 | |||
AK050358.1 | - | 19, 20 | 2514 | |||
AK051778.1 | - | 19, 20 | 2491 | |||
AK078183.1 | - | 19, 20 | 2533 | |||
AK085714.1 | - | 19, 20 | 2529 | |||
AK088965.1 | - | 19, 20 | 2492 | |||
AK166832.1 | - | 19, 20 | 2514 | |||
BC027432.1 | - | 9, 10 | 989 | |||
BC049238.1 | - | 3, 4 | 270 | |||
BC052403.1 | - | 19, 20 | 2508 | 5715723 |