Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198653.2 | NP_941055.1 | 5 | 834 | ||
GenBank mRNA | AK036182.1 | - | 5 | 802 | ||
AK043590.1 | - | 5 | 836 | |||
AK043729.1 | - | 5 | 812 | |||
AK048653.1 | - | 5 | 835 | |||
AK050358.1 | - | 5 | 817 | |||
AK051778.1 | - | 5 | 795 | |||
AK078183.1 | - | 5 | 836 | |||
AK085714.1 | - | 5 | 831 | |||
AK088965.1 | - | 5 | 795 | |||
AK164435.1 | - | 4 | 1217 | |||
AK166832.1 | - | 5 | 817 | |||
BC052403.1 | - | 5 | 811 | 5715723 |